Skip to Content
Merck
Tris(2,2,2-trifluoroethyl) phosphite 99%

Tris(2,2,2-trifluoroethyl) phosphite

Tris(2,2,2-trifluoroethyl) phosphite 99%
Linear Formula:
(CF3CH2O)3P
CAS Number:
Molecular Weight:
328.07

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing