Skip to Content
Merck
Tris(1,1,1,3,3,3-hexafluoro-2-propyl) phosphite 98%

Tris(1,1,1,3,3,3-hexafluoro-2-propyl) phosphite

Tris(1,1,1,3,3,3-hexafluoro-2-propyl) phosphite 98%
Synonym(s):
Tris(2H-perfluoro-2-propyl) phosphite
Linear Formula:
[(CF3)2CHO]3P
CAS Number:
Molecular Weight:
532.06

Products

Product Category
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing