Skip to Content
Merck
Trimethylsulfonium bromide 97%

Trimethylsulfonium bromide

Trimethylsulfonium bromide 97%
Empirical Formula (Hill Notation):
C3H9BrS
CAS Number:
Molecular Weight:
157.07

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing