![Thiamine Impurity E Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/148/866/a0ddabc9-0667-4636-8507-4cc8668476a2/640/a0ddabc9-0667-4636-8507-4cc8668476a2.jpg)
Thiamine Impurity E
![Thiamine Impurity E Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/148/866/a0ddabc9-0667-4636-8507-4cc8668476a2/640/a0ddabc9-0667-4636-8507-4cc8668476a2.jpg)
Synonym(s):
3-[(4-Amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methylthiazol-2(3H)-thione
Empirical Formula (Hill Notation):
C12H16N4OS2
CAS Number:
Molecular Weight:
296.41
Products
Brand
Formula Weight
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing