
Tetrahydrofuran-d8

Synonym(s):
Deuterated tetrahydrofuran, Octadeuterotetrahydrofuran
Empirical Formula (Hill Notation):
C4D8O
CAS Number:
Molecular Weight:
80.16
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Isotope
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing