Skip to Content
Merck
Sodium metavanadate anhydrous, 99.9% trace metals basis

Sodium metavanadate

Sodium metavanadate anhydrous, 99.9% trace metals basis
Synonym(s):
Sodium (meta)vanadate, Sodium trioxovanadate, Sodium vanadate(V), Sodium vanadium oxide, Sodium vanadium trioxide
Linear Formula:
NaVO3
CAS Number:
Molecular Weight:
121.93
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Reagent Type
Core
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing