
Sinensetin

Synonym(s):
2-(3,4-Dimethoxyphenyl)-5,6,7-trimethoxy-4H-1-benzopyran-4-one, 2-(3,4-Dimethoxyphenyl)-5,6,7-trimethoxychromen-4-one, 3′,4′,5,6,7-Pentamethoxyflavone, 6-Hydroxyluteolin 5,6,7,3′,4′-pentamethyl ether, Pedalitin permethyl ether
Empirical Formula (Hill Notation):
C20H20O7
CAS Number:
Molecular Weight:
372.37
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing