
Schisandrin A

Synonym(s):
Schisandrin A, Deoxyschisandrin, Deoxyschizandrin, Schizandrin A
Empirical Formula (Hill Notation):
C24H32O6
CAS Number:
Molecular Weight:
416.51
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing