
Sakuranetin

Synonym(s):
4′-5-Dihydroxy-7-methoxyflavanone, 7-O-Methylnaringenin, Naringenin 7-O-methyl ether
Empirical Formula (Hill Notation):
C16H14O5
CAS Number:
Molecular Weight:
286.28
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing