
Rimsulfuron

Synonym(s):
1-(4,6-Dimethoxy-2-pyrimidinyl)-3-[3-(ethylsulfonyl)-2-pyridylsulfonyl]urea
Empirical Formula (Hill Notation):
C14H17N5O7S2
CAS Number:
Molecular Weight:
431.44
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2