
Rimantadine hydrochloride

Synonym(s):
1-(1-Adamantyl)ethylamine hydrochloride, α-Methyltricyclo[3.3.1.13,7]-decane-1-methanamine hydrochloride, Rimantadine hydrochloride
Empirical Formula (Hill Notation):
C12H21N · HCl
CAS Number:
Molecular Weight:
215.76
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2