
Progesterone for peak identification

Synonym(s):
Progesterone, 4-Pregnene-3,20-dione
Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing