
Potassium tetraoxalate dihydrate

Synonym(s):
Potassium tetraoxalate dihydrate, Oxalic acid hemipotassium salt
Linear Formula:
KH3(C2O4)2 · 2H2O
CAS Number:
Molecular Weight:
254.19
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing