
pentanamide (valeramide)

Synonym(s):
Valeramide, NSC 400223, Pentanamide, Valeric acid amide
Empirical Formula (Hill Notation):
C5H11NO
CAS Number:
Molecular Weight:
101.15
Products
Brand
Formula Weight
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing