
N-norverapamil

Synonym(s):
(±)-Norverapamil hydrochloride, N-Nor-(±)-verapamil hydrochloride
Empirical Formula (Hill Notation):
C26H36N2O4 · HCl
CAS Number:
Molecular Weight:
477.04
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing