
Mesotrion Metabolite AMBA

Synonym(s):
2-Amino-4-(methylsulfonyl)benzoic acid
Empirical Formula (Hill Notation):
C8H9NO4S
CAS Number:
Molecular Weight:
215.23
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing