
Mercury(II) bromide

Synonym(s):
Mercuric bromide, Mercury(II) bromide
Linear Formula:
HgBr2
CAS Number:
Molecular Weight:
360.40
Products
Product Category
Brand
Boiling Point (°C)
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Reagent Type
Core
Special Grade
Showing 1-3 of 3