
Lysine hydrochloride

Synonym(s):
L-Lysine monohydrochloride, (S)-2,6-Diaminohexanoic acid monohydrochloride
Linear Formula:
H2N(CH2)4CH(NH2)CO2H·HCl
CAS Number:
Molecular Weight:
182.65
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing