
Losartan Related Compound B

Synonym(s):
2-Butyl-4-chloro-1-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-1H-imidazole-5-methanol 5-acetate, 2-Butyl-4-chloro-(1-{[2′-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl}-1H-imidazol-5-yl)methyl acetate
Empirical Formula (Hill Notation):
C24H25ClN6O2
CAS Number:
Molecular Weight:
464.95
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2