Skip to Content
Merck
L-Histidine-13C6 hydrochloride monohydrate 97 atom % 13C, 98% (CP)

L-Histidine-13C6 hydrochloride monohydrate

L-Histidine-13C6 hydrochloride monohydrate 97 atom % 13C, 98% (CP)
Synonym(s):
L-α-Amino-β-(4-imidazolyl)propionic acid-13C6 hydrochloride monohydrate
Empirical Formula (Hill Notation):
13C6H9N3O2HClH2O
CAS Number:
Molecular Weight:
215.59

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing