Skip to Content
Merck
Iodosulfuron-methyl-sodium PESTANAL®, analytical standard

Iodosulfuron-methyl-sodium

Iodosulfuron-methyl-sodium PESTANAL®, analytical standard
Synonym(s):
Methyl 4-iodo-2-[3-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)ureidosulfonyl]benzoate sodium salt
Empirical Formula (Hill Notation):
C14H13IN5NaO6S
CAS Number:
Molecular Weight:
529.24

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique