Skip to Content
Merck
Hydrocortisone butyrate United States Pharmacopeia (USP) Reference Standard

Hydrocortisone butyrate

Hydrocortisone butyrate United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Hydrocortisone 17-butyrate, 11β,17,21-Trihydroxypregn-4-ene-3,20-dione 17-butyrate, Cortisol 17-butyrate
Empirical Formula (Hill Notation):
C25H36O6
CAS Number:
Molecular Weight:
432.55

Products

Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing