
Hulupinic acid

Synonym(s):
4,5-Dihydroxy-2,2-bis(3-methyl-2-butenyl)- 4-cyclopentene-1,3-dione
Empirical Formula (Hill Notation):
C15H20O4
CAS Number:
Molecular Weight:
264.32
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing