
Glimepiride for system suitability

Synonym(s):
Glimepiride
Empirical Formula (Hill Notation):
C24H34N4O5S
CAS Number:
Molecular Weight:
490.62
Products
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2