
Folpet

Synonym(s):
2-(Trichloromethylsulfanyl)isoindole-1,3-dione
Empirical Formula (Hill Notation):
C9H4Cl3NO2S
CAS Number:
Molecular Weight:
296.56
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2