
Eupatorin

Synonym(s):
3′,5-Dihydroxy-4′,6,7-trimethoxyflavone, 5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-6,7-dimethoxy-4H-1-benzopyran-4-one, 6-Methoxyluteolin 4′,7-dimethyl ether, NSC 106402
Empirical Formula (Hill Notation):
C18H16O7
CAS Number:
Molecular Weight:
344.32
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Quality Segment
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing