
Ethyl-2-(4-hydroxy-3-methoxyphenyl)acetate

Synonym(s):
Ethyl homovanillate, Ethyl 4-hydroxy-3-methoxyphenylacetate
Linear Formula:
HOC6H3(OCH3)CH2CO2C2H5
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing