![Doxycycline hyclate British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/219/415/0ff6a5d1-3355-4e2f-8322-697cba5184bf/640/0ff6a5d1-3355-4e2f-8322-697cba5184bf.png)
Doxycycline hyclate
![Doxycycline hyclate British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/219/415/0ff6a5d1-3355-4e2f-8322-697cba5184bf/640/0ff6a5d1-3355-4e2f-8322-697cba5184bf.png)
Synonym(s):
Doxycycline hydrochloride hemiethanolate hemihydrate, Doxycycline hyclate
Empirical Formula (Hill Notation):
C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O
CAS Number:
Molecular Weight:
512.94
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing