
DMT-dG(tac) Phosphoramidite

Synonym(s):
DMT-dG(tac) amidite
Empirical Formula (Hill Notation):
C52H62N7O9P
CAS Number:
Molecular Weight:
960.06
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Showing 1-3 of 3