
DMT-dC(ac) Phosphoramidite

Synonym(s):
DMT-dC(ac) Amidite, N-acetyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxycytidine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite], N4-Acetyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxycytidine-3′-O-[O-(2-cyanoethyl)-N,N′-diisopropylphosphoramidite]
Empirical Formula (Hill Notation):
C41H50N5O8P
CAS Number:
Molecular Weight:
771.84
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing