
Dipivefrine hydrochloride

Synonym(s):
[2-(2,2-Dimethylpropanoyloxy)-4-[1-hydroxy-2-(methylamino)ethyl]phenyl] 2,2-dimethylpropanoate hydrochloride
Empirical Formula (Hill Notation):
C19H29NO5 · HCl
CAS Number:
Molecular Weight:
387.90
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Special Grade
Showing 1-2 of 2