
Diphenhydramine Related Compound A

Synonym(s):
2-(Diphenylmethoxy)-N-methylethanamine hydrochloride, N-Desmethyldiphenhydramine hydrochloride
Empirical Formula (Hill Notation):
C16H19NO · HCl
CAS Number:
Molecular Weight:
277.79
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2