![Dimepiperate PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/281/733/bfae6b82-e981-4af0-899a-b1b36b5c3ec5/640/bfae6b82-e981-4af0-899a-b1b36b5c3ec5.png)
Dimepiperate
![Dimepiperate PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/281/733/bfae6b82-e981-4af0-899a-b1b36b5c3ec5/640/bfae6b82-e981-4af0-899a-b1b36b5c3ec5.png)
Synonym(s):
S-(1-Methyl-1-phenylethyl)-1-piperidinecarbothioate
Empirical Formula (Hill Notation):
C15H21NOS
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing