
D-Glucose-1,6-13C2

Synonym(s):
Dextrose-1,6-13C2, Dextrose-1,6-13C2
Empirical Formula (Hill Notation):
13C2C4H12O6
CAS Number:
Molecular Weight:
182.14
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Showing 1-2 of 2
Compare
Product Number
Description
Pricing