
Daunorubicin hydrochloride

Synonym(s):
Daunomycin hydrochloride, Daunorubicin hydrochloride
Empirical Formula (Hill Notation):
C27H29NO10 · HCl
CAS Number:
Molecular Weight:
563.98
Products
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing