
Cytidine 2′:3′-cyclic monophosphate monosodium salt

Synonym(s):
2′,3′-Cyclic CMP monosodium salt, Cyclic cytidylic acid, Cytidine 2′,3′-cyclophosphate sodium salt
Empirical Formula (Hill Notation):
C9H11N3NaO7P
Products
Brand
Biological Source
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing