
Clofentezine

Synonym(s):
3,6-Bis(2-chlorophenyl)-1,2,4,5-tetrazine, Clofentezine
Empirical Formula (Hill Notation):
C14H8Cl2N4
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2