
Chlorhexidine dihydrochloride

Synonym(s):
1,6-Bis(N5-[p-chlorophenyl]-N1-biguanido)hexane; 1,1′-Hexamethylenebis(5-[p-chlorophenyl]biguanide), Lisium
Empirical Formula (Hill Notation):
C22H30Cl2N10 · 2HCl
CAS Number:
Molecular Weight:
578.37
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing