
Bakuchiol

Synonym(s):
(S)-Bakuchiol, 4-[(1E,3S)-3-Ethenyl-3,7-dimethyl-1,6-octadien-1-yl]phenol
Empirical Formula (Hill Notation):
C18H24O
CAS Number:
Molecular Weight:
256.38
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing