![Aripiprazole Related Compound B Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/399/395/ec26f655-ec09-4724-b18e-49a44e4b68ef/640/ec26f655-ec09-4724-b18e-49a44e4b68ef.jpg)
Aripiprazole Related Compound B
![Aripiprazole Related Compound B Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/399/395/ec26f655-ec09-4724-b18e-49a44e4b68ef/640/ec26f655-ec09-4724-b18e-49a44e4b68ef.jpg)
Synonym(s):
7-(4-Hydroxybutoxy)-3,4-dihydroquinolin-2(1H)-one
Empirical Formula (Hill Notation):
C13H17NO3
CAS Number:
Molecular Weight:
235.28
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2