
Anti-VSV-Glycoprotein−Agarose antibody, Mouse monoclonal

Synonym(s):
Monoclonal Anti-VSV Glycoprotein
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Analyte Suitability
Antibody Form
Conjugate
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing