
Anti-Nerve Growth Factor Receptor/TNFRSF16 antibody produced in goat

Synonym(s):
Anti-LNGFR, Anti-Ngfr, Anti-Tnfrsf16, Anti-p75
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing