
Anti-IL1RL1 antibody produced in rabbit

Synonym(s):
Anti-Interleukin-1 receptor-like 1 precursor antibody produced in rabbit, Anti-ST2 protein antibody produced in rabbit, Anti-DER4, Anti-FIT-1, Anti-Interleukin 1 receptor-like 1, Anti-MGC32623, Anti-ST2
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing