
Acridine Orange 10-nonyl bromide

Synonym(s):
10-Nonylacridine orange bromide, 3,6-Bis(dimethylamino)-10-nonylacridinium bromide
Empirical Formula (Hill Notation):
C26H38BrN3
CAS Number:
Molecular Weight:
472.50
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing