Skip to Content
Merck
Acitretin for impurity A identification European Pharmacopoeia (EP) Reference Standard

Acitretin for impurity A identification

Acitretin for impurity A identification European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Acitretin, 9-(4-Methoxy-2,3,6-trimethylphenyl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid
Empirical Formula (Hill Notation):
C21H26O3
CAS Number:
Molecular Weight:
326.43
EC Number:

Products

Formula Weight
Gene Alias
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing