
α,α-Diphenyl-γ-butyrolactone

Synonym(s):
2,2-Diphenylbutyrolactone, 4-Hydroxy-2,2-diphenylbutyric acid γ-lactone, Dihydro-3,3-diphenyl-2(3H)-furanone
Empirical Formula (Hill Notation):
C16H14O2
CAS Number:
Molecular Weight:
238.28
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing