5-(Iodoacetamido)fluorescein
Synonym(s):
5-IAF
Empirical Formula (Hill Notation):
C22H14INO6
CAS Number:
Molecular Weight:
515.25
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing