Skip to Content
Merck
4-Bromo-2-thiophenecarboxaldehyde 90%

4-Bromo-2-thiophenecarboxaldehyde

4-Bromo-2-thiophenecarboxaldehyde 90%
Empirical Formula (Hill Notation):
C5H3BrOS
CAS Number:
Molecular Weight:
191.05
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing