Skip to Content
Merck
4-(4-methoxyphenyl)benzaldehyde 97%

4-(4-methoxyphenyl)benzaldehyde

4-(4-methoxyphenyl)benzaldehyde 97%
Synonym(s):
4′-Methoxybiphenyl-4-carboxaldehyde
Empirical Formula (Hill Notation):
C14H12O2
CAS Number:
Molecular Weight:
212.24

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing