
2-Methylbutyryl-L-carnitine

Synonym(s):
(2R)-3-Carboxy-N,N,N-trimethyl-2-(2-methyl-1-oxobutoxy)-1-propanaminium inner salt
Empirical Formula (Hill Notation):
C12H23NO4
CAS Number:
Molecular Weight:
245.32
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing